missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucuronic acid γ-lactone, ≥99% (sum of anomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111013923
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013923 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013923 Supplier Merck Emd Millipore Supplier No. 05566100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

D-Glucuronic acid γ-lactone is a normal human metabolite produced from glucose as starting material. It serves as a precursor for the synthesis of a number of deoxy sugars.

Specifications

CAS 32449-92-6
Linear Formula C6H8O6
MDL Number MFCD00135622
Melting Point 172°C to 175°C
Synonym D-(+)-Glucurono-6,3-lactone; D-Glucurone; D-Glucurono-6,3-lactone; Glucuronolactone
Type Analytical Standard
Quantity 100 mg
Formula Weight 176.12
Percent Purity ≥99% (sum of anomers, GC)
Grade Analytical Standard