missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucose 98.0+%, TCI America™

Catalog No. G004825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G004825G 25 g
G0048500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G004825G Supplier TCI America Supplier No. G004825G
Only null left

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00148912
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 146°C
Color White
Quantity 25 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Glucose
EINECSNumber (8)-0046
RTECSNumber LZ6600000
TSCA Yes