missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucose Solution, MilliporeSigma™ Supelco™

Catalog No. 111014391
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014391 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014391 Supplier Merck Emd Millipore Supplier No. G3285.5ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Food and Beverages and General Analytical

Specifications

CAS 50-99-7
Purity Grade Notes Traceable to a NIST standard
Concentration 1 mg/mL in 0.1% benzoic acid
Linear Formula C6H12O6
MDL Number MFCD00063774
Type Analytical Standard
Quantity 0.5 mL
Formula Weight 180.16
Grade Analytical Standard