missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucose, Molecular biology reagent grade, MP Biomedicals™

Catalog No. ICN19402491
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19402491 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19402491 Supplier MP Biomedicals Inc Supplier No. 0219402491
Only null left

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Color White
Density 1.544g/mL
Quantity 1 kg
Formula Weight 180.2
Grade Molecular Biology Reagent
Physical Form Granular Powder
Chemical Name or Material D-(+)-Glucose
EINECSNumber 200-075-1
Recommended Storage Store at room temperature.