missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Glucose dehydrogenase

Catalog No. AA454024I
Change view
Click to view available options
Quantity:
1 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA454024I 1 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA454024I Supplier Thermo Scientific Chemicals Supplier No. 0454024I
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9028-53-9

Diagnostics and cofactor regeneration of NAD(P)H in reductive processes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9028-53-9
MDL Number MFCD00131181

Specifications

Color Yellow
Quantity 1 kU
Physical Form Powder
Chemical Name or Material D-Glucose dehydrogenase
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only