CAS RN 9028-53-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals D-Glucose dehydrogenase
Thermo Scientific Chemicals D-Glucose dehydrogenase
CAS: 9028-53-9 MDL Number: MFCD00131181