missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP

Catalog No. AC388190250
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC388190250 25 kg
AC388190010 1 kg
AC388190050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC388190250 Supplier Thermo Scientific Chemicals Supplier No. 388190250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50-99-7 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 146.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 16 h)
Quantity 25 kg
Merck Index 15,4495
Specific Rotation Condition + 52.75 (25.00°C c=10, H2O, NH4OH)
Specific Rotation + 52.75
Solubility Information Solubility in water: soluble.
Formula Weight 180.16
Percent Purity 97.5 to 102.0% (Anhydrous substance)
Related Substances 0.5% max. (total impurities)
Chemical Name or Material D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP
Show More Show Less
EINECSNumber 200-075-1

RUO – Research Use Only