missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Glucose, anhydrous, For ACS analysis, MP Biomedicals™
Specifications
Specifications
| Density | 1.544g/mL |
| Quantity | 500 g |
| Formula Weight | 180.2 |
| Grade | ACS |
| Physical Form | Powder |
| Chemical Name or Material | D-(+)-Glucose, For ACS analysis |