missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Glucose, anhydrous, 99%

CAS: 50-99-7 | C6H12O6 | 180.156 g/mol

Supplier:  Thermo Scientific Chemicals A168280E

 View more versions of this product

Catalog No. AAA168280E


Only null left

Description

Description

D-Glucose is used in various metabolic processes including enzymic synthesis of cyclohexyl-alpha and beta -D-glucosides. It is involved in the detection of type 2 diabetes mellitus and the most widely used aldohexose in living organisms. Its availability influences psychological processes since it is a primary energy source for the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glucose is used in various metabolic processes including enzymic synthesis of cyclohexyl-alpha and beta -D-glucosides. It is involved in the detection of type 2 diabetes mellitus and the most widely used aldohexose in living organisms. Its availability influences psychological processes since it is a primary energy source for the brain.

Solubility
Soluble in water, hot glacial acetic acid, pyridine, aniline and dimethyl sulfoxide. Slightly soluble in methanol, ethanol and acetone.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

D-(+)-Glucose
50-99-7
99%
MFCD00063774
1281608
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
GZCGUPFRVQAUEE-SLPGGIOYSA-N
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
107526
180.16
anhydrous
147°C to 153°C
C6H12O6
2500 g
14,4459
Soluble in water,hot glacial acetic acid,pyridine,aniline and dimethyl sulfoxide. Slightly soluble in methanol,ethanol and acetone.
C(C(C(C(C(C=O)O)O)O)O)O
180.156
CHEBI:42758
99%

Chemical Identifiers

50-99-7
180.156
GZCGUPFRVQAUEE-SLPGGIOYSA-N
107526
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
C6H12O6
MFCD00063774
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
CHEBI:42758
C(C(C(C(C(C=O)O)O)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-075-1

RTECSNumber : LZ6600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only