missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Glucose, anhydrous, 99%

Catalog No. AAA1682836
Change view
Click to view available options
Quantity:
10,000 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1682836 500 g
AAA168280E 2500 g
AAA168280C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1682836 Supplier Thermo Scientific Chemicals Supplier No. A1682836

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50-99-7 | C6H12O6 | 180.156 g/mol

D-Glucose is used in various metabolic processes including enzymic synthesis of cyclohexyl-alpha and beta -D-glucosides. It is involved in the detection of type 2 diabetes mellitus and the most widely used aldohexose in living organisms. Its availability influences psychological processes since it is a primary energy source for the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glucose is used in various metabolic processes including enzymic synthesis of cyclohexyl-alpha and beta -D-glucosides. It is involved in the detection of type 2 diabetes mellitus and the most widely used aldohexose in living organisms. Its availability influences psychological processes since it is a primary energy source for the brain.

Solubility
Soluble in water, hot glacial acetic acid, pyridine, aniline and dimethyl sulfoxide. Slightly soluble in methanol, ethanol and acetone.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00063774
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 147°C to 153°C
Assay Percent Range 99%
Quantity 500 g
Beilstein 1281608
Merck Index 14,4459
Solubility Information Soluble in water,hot glacial acetic acid,pyridine,aniline and dimethyl sulfoxide. Slightly soluble in methanol,ethanol and acetone.
Formula Weight 180.16
Percent Purity 99%
Chemical Name or Material D-(+)-Glucose, anhydrous
EINECSNumber 200-075-1
RTECSNumber LZ6600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only