missing translation for 'onlineSavingsMsg'
Learn More

D-Glucosamine sulfate, MP Biomedicals™

Catalog No. p-4679618
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15392810 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15392810 Supplier MP Biomedicals Inc Supplier No. 0215392810

Chemical Identifiers

CAS 29031-19-4
Molecular Formula C6H15NO9S
Molecular Weight (g/mol) 277.244
InChI Key FGNPLIQZJCYWLE-BTVCFUMJSA-N
Synonym d-glucosamine sulphate, 2r,3r,4s,5r-2-amino-3,4,5,6-tetrahydroxyhexanal sulfate, gevolox, glucosaminsulfat, sulfato de glucosamina, 2-amino-2-deoxy-d-glucose sulfate salt, glucosamine sulfate 2kcl, d-glucose, 2-amino-2-deoxy-, sulfate salt, glucosamine sulfate na salt, glucosamine sulfate dc
PubChem CID 115046
IUPAC Name (2R,3R,4S,5R)-2-amino-3,4,5,6-tetrahydroxyhexanal;sulfuric acid
SMILES C(C(C(C(C(C=O)N)O)O)O)O.OS(=O)(=O)O

Specifications

Quantity 10 g
Formula Weight 228.2
Physical Form Solid
Chemical Name or Material D-Glucosamine Sulfate
EINECSNumber 249-379-6
Recommended Storage Store at room temperature.