accessibility menu, dialog, popup

UserName

NH2(R)O(R)OH(S)OH(R)OHOHOHSOHOO

CAS RN 29031-19-4

IUPAC Name: (2R,3R,4S,5R)-2-amino-3,4,5,6-tetrahydroxyhexanal; sulfuric acid
Synonyms: glucosamine sulfate
Molecular Weight (g/mol): 277.24
Molecular Formula: C6H15NO9S
InChi Key: FGNPLIQZJCYWLE-BTVCFUMJSA-N
SMILES: OS(O)(=O)=O.N[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results