missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Glucosamine Hydrochloride 98.0+%, TCI America™

$52.92 - $292.92
Chemical Identifiers
CAS | 66-84-2 |
---|---|
Molecular Formula | C6H13NO5 |
MDL Number | MFCD00135831 |
Synonym | 2-Amino-2-deoxy-D-glucopyranose Hydrochloride |
Chemical Identifiers
66-84-2 | |
C6H13NO5 | |
MFCD00135831 | |
2-Amino-2-deoxy-D-glucopyranose Hydrochloride |
Specifications
66-84-2 | |
White | |
MFCD00135831 | |
2-Amino-2-deoxy-D-glucopyranose Hydrochloride | |
≥98.0% (T) | |
D-(+)-Glucosamine Hydrochloride |
194°C | |
C6H13NO5 | |
25 g | |
215.63 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : LZ6665000
TSCA : Yes