missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucosamine Hydrochloride 98.0+%, TCI America™

Catalog No. G0044500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0044500G 500 g
G004425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0044500G Supplier TCI America Supplier No. G0044500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66-84-2
Molecular Formula C6H13NO5
MDL Number MFCD00135831
Synonym 2-Amino-2-deoxy-D-glucopyranose Hydrochloride

Specifications

Melting Point 194°C
Color White
Quantity 500 g
Formula Weight 215.63
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Glucosamine Hydrochloride
RTECSNumber LZ6665000
TSCA Yes