Learn More
Thermo Scientific Chemicals D-Glucosamine hydrochloride, 98+%
D-Glucosamine hydrochloride, CAS # 3416-24-8, is the salt of the amino sugar D-Glucosamine, used for chitin quantitation and other biochemical study.
$73.42 - $700.82
Chemical Identifiers
| CAS | 66-84-2 |
|---|---|
| Molecular Formula | C6H13NO5·HCl |
| MDL Number | MFCD00067674 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1553218
|
Thermo Scientific Chemicals
A1553218 |
50 g |
Each for $73.42
|
|
|||||
|
AAA1553230
|
Thermo Scientific Chemicals
A1553230 |
250 g |
Each for $197.71
|
|
|||||
|
AAA155320B
|
Thermo Scientific Chemicals
A155320B |
1000 g |
Each for $700.82
|
|
|||||
Description
Novel application of glucosamine is to prepare medical agent for treating vertigo. It is used as food ingredients and additives, raw material for anti cancer and antibiotic drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• D-Glucosamine is an abundant monosaccharide amino sugar that acts as a building block in the synthesis of glycosylated proteins and lipids
• D-Glucosamine is the precursor at the start of the hexosamine pathway that is essential to the formation of common proteoglycans
• D-Glucosamine combines with chondroitin sulfate in cartilage and is found as a monomer in chitin
Application
• D-Glucosamine can be used as a biochemical standard or for cell stimulation along with glucose in cell culture
• It may be useful in the laboratory study of osteoarthritis and diabetes
Chemical Identifiers
| 66-84-2 | |
| C6H13NO5·HCl | |
| MFCD00067674 |
Specifications
| 66-84-2 | |
| C6H13NO5·HCl | |
| 50 g | |
| 14,4458 | |
| +71.5° (c=1 in Water, 20h) | |
| ≥98% |
| ∼192°C (decomposition) | |
| MFCD00067674 | |
| 4157370 | |
| Soluble in water. | |
| 215.64 | |
| D-Glucosamine hydrochloride |
Safety and Handling
EINECSNumber : 200-638-1
RTECSNumber : LZ6665000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only