missing translation for 'onlineSavingsMsg'
Learn More

D-Glucamine 97.0+%, TCI America™

Catalog No. G02525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02525G 5 g
G025225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02525G Supplier TCI America Supplier No. G02525G

Chemical Identifiers

CAS 488-43-7
Molecular Formula C6H15NO5
Molecular Weight (g/mol) 181.19
MDL Number MFCD00077776
InChI Key SDOFMBGMRVAJNF-UHFFFAOYNA-N
Synonym 1-Amino-1-deoxy-D-glucitol, 1-Amino-1-deoxy-D-sorbitol
PubChem CID 452501
IUPAC Name 6-aminohexane-1,2,3,4,5-pentol
SMILES NCC(O)C(O)C(O)C(O)CO

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 181.19
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Glucamine