missing translation for 'onlineSavingsMsg'
Learn More

D-Glucamine 97.0+%, TCI America™

Supplier:  TCI America G025225G

Quantity 25 g

 View more versions of this product

Catalog No. G025225G

Only null left

Description

Description

Specifications

Chemical Identifiers

488-43-7
181.19
SDOFMBGMRVAJNF-UHFFFAOYNA-N
452501
NCC(O)C(O)C(O)C(O)CO
C6H15NO5
MFCD00077776
1-Amino-1-deoxy-D-glucitol, 1-Amino-1-deoxy-D-sorbitol
6-aminohexane-1,2,3,4,5-pentol

Specifications

D-Glucamine
White-Yellow
25 g
1-Amino-1-deoxy-D-glucitol, 1-Amino-1-deoxy-D-sorbitol
NCC(O)C(O)C(O)C(O)CO
181.19
181.19
Crystalline Powder
488-43-7
C6H15NO5
MFCD00077776
SDOFMBGMRVAJNF-UHFFFAOYNA-N
6-aminohexane-1,2,3,4,5-pentol
452501
≥97.0% (T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions