missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Galacturonic acid monohydrate, ≥95.0% (TLC); 97.0-101.0% (T), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 91510-62-2 |
| Linear Formula | C6H10O7 · H2O |
| MDL Number | MFCD00071585 |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 212.15 |
| Percent Purity | ≥95.0% (TLC); 97.0 to 101.0% (T) |
| Grade | Analytical Standard |