missing translation for 'onlineSavingsMsg'
Learn More

D-Galactose, Anhydrous, Reagent, 99%, Spectrum™ Chemical

Catalog No. 18602123
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602123 500 g Poly Bottle
18602122 100 g Poly Bottle
18602124 1 kg Poly Bottle
18602125 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602123 Supplier Spectrum Chemical Mfg Cor Supplier No. GA110500GM
Only null left

GA110, 59-23-4, C6H12O6

Spectrum™ Chemical D-Galactose, Anhydrous, Reagent

Chemical Identifiers

CAS 59-23-4
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
IUPAC Name 6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OCC1OC(O)C(O)C(O)C1O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99%
Grade Reagent