missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Galactose, 99+%

Catalog No. p-3736550
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC150611000 100 g
AC150615000 500 g
AC150610010 1 kg
AC150610051 5 kg
AC150610250 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC150611000 Supplier Thermo Scientific Chemicals Supplier No. 150611000
Only null left

D-Galactose, CAS # 59-23-4, is a right-handed conformation of a monosaccharide that is grouped as an aldohexose, and the molecule may be useful in study of aging, inflammation, and metabolic disorders. | CAS: 59-23-4 | C6H12O6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Galactose is a monosaccharide and aldohexose found in most organisms
  • Galactose is an epimer of glucose and can be converted to glucose through the Leloir pathway
  • It also plays a role as a metabolite in mice and E. coli
  • Galactose is less stable than glucose and rapid conversion often occurs
  • Galactose may also be incorporated into glycoconjugates

Applications

  • Metabolism of galactose may be relevant to the study of aging, inflammation, and metabolic disorders
  • It is intended for a variety of applications from in vitro metabolic assays to HPLC

Chemical Identifiers

CAS 59-23-4
Molecular Formula C6H12O6
MDL Number MFCD00063833

Specifications

Melting Point 165.0°C to 168.0°C
Color White
Density 6.2
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 100 g
Beilstein 31, 295
Merck Index 15, 4365
Specific Rotation Condition + 79.75 (20.00°C c=10,H2O,NH3)
Specific Rotation + 79.75
Vapor Pressure Negligible
Formula Weight 180.16
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material D(+)-Galactose
Show More Show Less

RUO – Research Use Only