missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Galactal 98.0+%, TCI America™
$62.08 - $62.08
Chemical Identifiers
| CAS | 21193-75-9 |
|---|---|
| Molecular Formula | C6H10O4 |
| Molecular Weight (g/mol) | 146.142 |
| MDL Number | MFCD00038067 |
| InChI Key | YVECGMZCTULTIS-HSUXUTPPSA-N |
| Synonym | d-galactal, galactal, d-+-galactal, 1,5-anhydro-2-deoxy-d-lyxo-hex-1-enitol, 2r,3r,4r-2-hydroxymethyl-3,4-dihydro-2h-pyran-3,4-diol, arabino-hex-5-enitol, 2,6-anhydro-5-deoxy, 2,6-anhydro-5-deoxy-d-arabino-hex-5-enitol, 1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, pubchem10609 |
| PubChem CID | 2734735 |
| ChEBI | CHEBI:63139 |
| IUPAC Name | (2R,3R,4R)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol |
| SMILES | C1=COC(C(C1O)O)CO |
Chemical Identifiers
| 21193-75-9 | |
| 146.142 | |
| YVECGMZCTULTIS-HSUXUTPPSA-N | |
| 2734735 | |
| (2R,3R,4R)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol |
| C6H10O4 | |
| MFCD00038067 | |
| d-galactal, galactal, d-+-galactal, 1,5-anhydro-2-deoxy-d-lyxo-hex-1-enitol, 2r,3r,4r-2-hydroxymethyl-3,4-dihydro-2h-pyran-3,4-diol, arabino-hex-5-enitol, 2,6-anhydro-5-deoxy, 2,6-anhydro-5-deoxy-d-arabino-hex-5-enitol, 1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, pubchem10609 | |
| CHEBI:63139 | |
| C1=COC(C(C1O)O)CO |
Specifications
| 21193-75-9 | |
| MFCD00038067 | |
| d-galactal, galactal, d-+-galactal, 1,5-anhydro-2-deoxy-d-lyxo-hex-1-enitol, 2r,3r,4r-2-hydroxymethyl-3,4-dihydro-2h-pyran-3,4-diol, arabino-hex-5-enitol, 2,6-anhydro-5-deoxy, 2,6-anhydro-5-deoxy-d-arabino-hex-5-enitol, 1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, pubchem10609 | |
| C1=COC(C(C1O)O)CO | |
| 146.142 | |
| CHEBI:63139 | |
| ≥98.0% (HPLC) | |
| D-Galactal |
| C6H10O4 | |
| 1 g | |
| YVECGMZCTULTIS-HSUXUTPPSA-N | |
| (2R,3R,4R)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol | |
| 2734735 | |
| 146.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator