accessibility menu, dialog, popup

UserName

CAS RN 21193-75-9

HOOHOHO

CAS RN 21193-75-9

IUPAC Name: 2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol
Synonyms: 2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: YVECGMZCTULTIS-UHFFFAOYNA-N
SMILES: OCC1OC=CC(O)C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results