missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Fucose, 98+%

Catalog No. p-3758062
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC225630050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC225630050 Supplier Thermo Scientific Chemicals Supplier No. 225630050

D (+)-Fucose, 98+%, CAS # 3615-37-0, also known as aldehyde-D-fucose, is an enantiomer of an aldehyde-L-fucose found in plants. It shows several biological activities, including cellular adhesion. | CAS: 3615-37-0 | C6H12O5 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• D (+)-Fucose can be found mainly in plants and shows several biological activities, including cellular adhesion

Applications

• D-Fucose is usually found in plants and some microorganisms, as a relatively simple glycoside comprising only a few sugar units

• It can be used as a sole carbon source in some microorganisms

• It may be used to study enzymes involved in its mutarotation to form L-fucose

• This compound is used as a reference compound in the analysis of carbohydrate metabolites

Chemical Identifiers

CAS 3615-37-0
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00135603
InChI Key PNNNRSAQSRJVSB-DPYQTVNSSA-N
Synonym 2r,3s,4s,5r-2,3,4,5-tetrahydroxyhexanal, aldehydo-d-fucose, d +-fucose, galactose, 6-deoxy, d-galactose, 6-deoxy, fucose, d, +-fucose, d0o8ie, d-+-fucose, d-+-fucose;6-deoxy-d-galactose
PubChem CID 94270
ChEBI CHEBI:48203
IUPAC Name (2R,3S,4S,5R)-2,3,4,5-tetrahydroxyhexanal
SMILES CC(C(C(C(C=O)O)O)O)O

Specifications

Melting Point 144.0°C to 145.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 31, 76
Merck Index 15, 4307
Specific Rotation Condition + 76.00 (20.00°C c=4,H2O,NH3)
Specific Rotation + 76.00
Solubility Information (5% in water) Clear colorless to slightly yellow
Formula Weight 164.16
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Fucose
Show More Show Less
EINECSNumber 222-792-9

RUO – Research Use Only