missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Fucose, 98+%

D (+)-Fucose, 98+%, CAS # 3615-37-0, also known as aldehyde-D-fucose, is an enantiomer of an aldehyde-L-fucose found in plants. It shows several biological activities, including cellular adhesion. | CAS: 3615-37-0 | C6H12O5 | 164.16 g/mol

Supplier:  Thermo Scientific Chemicals 225630050

 View more versions of this product

Catalog No. AC225630050


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• D (+)-Fucose can be found mainly in plants and shows several biological activities, including cellular adhesion

Applications

• D-Fucose is usually found in plants and some microorganisms, as a relatively simple glycoside comprising only a few sugar units

• It can be used as a sole carbon source in some microorganisms

• It may be used to study enzymes involved in its mutarotation to form L-fucose

• This compound is used as a reference compound in the analysis of carbohydrate metabolites

Specifications

Specifications

D(+)-Fucose
144.0°C to 145.0°C
Authentic
Glass bottle
MFCD00135603
31, 76
+ 76.00 (20.00°C c=4,H2O,NH3)
2r,3s,4s,5r-2,3,4,5-tetrahydroxyhexanal, aldehydo-d-fucose, d +-fucose, galactose, 6-deoxy, d-galactose, 6-deoxy, fucose, d, +-fucose, d0o8ie, d-+-fucose, d-+-fucose;6-deoxy-d-galactose
PNNNRSAQSRJVSB-DPYQTVNSSA-N
(2R,3S,4S,5R)-2,3,4,5-tetrahydroxyhexanal
94270
164.16
Crystalline Powder
3615-37-0
White
98% min. (ELSD) (HPLC)
C6H12O5
5 g
15, 4307
+ 76.00
(5% in water) Clear colorless to slightly yellow
CC(C(C(C(C=O)O)O)O)O
164.16
CHEBI:48203
98+%

Chemical Identifiers

3615-37-0
164.16
PNNNRSAQSRJVSB-DPYQTVNSSA-N
94270
(2R,3S,4S,5R)-2,3,4,5-tetrahydroxyhexanal
C6H12O5
MFCD00135603
2r,3s,4s,5r-2,3,4,5-tetrahydroxyhexanal, aldehydo-d-fucose, d +-fucose, galactose, 6-deoxy, d-galactose, 6-deoxy, fucose, d, +-fucose, d0o8ie, d-+-fucose, d-+-fucose;6-deoxy-d-galactose
CHEBI:48203
CC(C(C(C(C=O)O)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 222-792-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only