missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Fructose, MilliporeSigma™, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014709
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014709 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014709 Supplier Merck Emd Millipore Supplier No. F2793500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

D-(−)-Fructose is a important constituent of sucrose, found in many plants storage organs, which contributes to fruit sweetness and is a major source of calories in fruit consumption.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 57-48-7
Physical Form Neat
Linear Formula C6H12O6
MDL Number MFCD00148910
Melting Point 119°C to 122°C
Synonym D-Levulose; Fruit sugar
Type Analytical Standard
Quantity 500 mg
Formula Weight 180.16
Percent Purity ≥99% (HPLC)
Grade Analytical Standard
Show More Show Less