missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Fructose, 99%

CAS: 57-48-7 | C6H12O6 | 180.156 g/mol

Supplier:  Thermo Scientific Chemicals A177180I

 View more versions of this product

Catalog No. AAA177180I

Only null left
Explore available promotions

Description

Description

Fructose is used commercially in foods and beverages. It increases starch viscosity more rapidly and achieves a higher final viscosity than sucrose. It has all the fiber squeezed out of it and so the fructose is easily absorbed into the bloodstream. High-fructose-corn syrup is used in frozen products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fructose is used commercially in foods and beverages. It increases starch viscosity more rapidly and achieves a higher final viscosity than sucrose. It has all the fiber squeezed out of it and so the fructose is easily absorbed into the bloodstream. High-fructose-corn syrup is used in frozen products.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

D-Fructose
∼105°C to 110°C
99%
C6H12O6
5000 g
14,4273
Soluble in water.
−89° (c=2 in Water)
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
5984
180.16
57-48-7
1.59
Odorless
MFCD00148910
1239004
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
BJHIKXHVCXFQLS-UYFOZJQFSA-N
C(C(C(C(C(=O)CO)O)O)O)O
180.156
CHEBI:48095
99%

Chemical Identifiers

57-48-7
180.156
BJHIKXHVCXFQLS-UYFOZJQFSA-N
5984
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
C6H12O6
MFCD00148910
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
CHEBI:48095
C(C(C(C(C(=O)CO)O)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-333-3

RTECSNumber : LS7120000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only