missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Fructose, 99%

Catalog No. p-3346072
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1771830 250 g
AAA177180B 1000 g
AAA177180I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1771830 Supplier Thermo Scientific Chemicals Supplier No. A1771830
Only null left

CAS: 57-48-7 | C6H12O6 | 180.156 g/mol

Fructose is used commercially in foods and beverages. It increases starch viscosity more rapidly and achieves a higher final viscosity than sucrose. It has all the fiber squeezed out of it and so the fructose is easily absorbed into the bloodstream. High-fructose-corn syrup is used in frozen products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fructose is used commercially in foods and beverages. It increases starch viscosity more rapidly and achieves a higher final viscosity than sucrose. It has all the fiber squeezed out of it and so the fructose is easily absorbed into the bloodstream. High-fructose-corn syrup is used in frozen products.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00148910
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
Synonym d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point ∽105°C to 110°C
Density 1.59
Assay Percent Range 99%
Quantity 250 g
Beilstein 1239004
Merck Index 14,4273
Solubility Information Soluble in water.
Formula Weight 180.16
Percent Purity 99%
Chemical Name or Material D-Fructose
EINECSNumber 200-333-3
RTECSNumber LS7120000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only