missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Fructose-6-phosphate disodium salt, 95%

CAS: 26177-86-6 | C6H13Na2O9P | 306.11 g/mol

$76.79 - $278.92

Chemical Identifiers

CAS 26177-86-6
Molecular Formula C6H13Na2O9P
Molecular Weight (g/mol) 306.11
MDL Number MFCD16661244
InChI Key VAPLWAFNCWDFQF-UHFFFAOYNA-N
PubChem CID 129893509
IUPAC Name sodium;[(2R,3R,4S)-2,3,4,6-tetrahydroxy-5-oxohexyl] dihydrogen phosphate
SMILES [Na].[Na].OCC(=O)C(O)C(O)C(O)COP(O)(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66311MC
View Documents
Thermo Scientific Chemicals
J66311MC
100 mg
Each for $76.79
Only null left
 
AAJ66311ME
View Documents
Thermo Scientific Chemicals
J66311ME
500 mg
Each for $168.42
Only null left
 
AAJ6631103
View Documents
Thermo Scientific Chemicals
J6631103
1 g
Each for $278.92
Only null left
 
Description

Description

D-Fructose-6-phosphate disodium salt is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amidotransferase(s) and glucosamine-6P synthase(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-6-phosphate disodium salt is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amidotransferase(s) and glucosamine-6P synthase(s).

Solubility
Soluble in water (100 mg/ml).

Notes
Store at -20°C. Incompatible with heat and oxidizing agents.
Specifications

Chemical Identifiers

26177-86-6
306.11
VAPLWAFNCWDFQF-UHFFFAOYNA-N
sodium;[(2R,3R,4S)-2,3,4,6-tetrahydroxy-5-oxohexyl] dihydrogen phosphate
C6H13Na2O9P
MFCD16661244
129893509
[Na].[Na].OCC(=O)C(O)C(O)C(O)COP(O)(O)=O

Specifications

26177-86-6
C6H13Na2O9P
100 mg
VAPLWAFNCWDFQF-UHFFFAOYNA-N
sodium;[(2R,3R,4S)-2,3,4,6-tetrahydroxy-5-oxohexyl] dihydrogen phosphate
129893509
95%
D-Fructose-6-phosphate disodium salt
White
MFCD16661244
Soluble in water (100mg/ml).
[Na].[Na].OCC(=O)C(O)C(O)C(O)COP(O)(O)=O
306.11
304.1
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 247-505-4

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only