missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Fructose-6-phosphate disodium salt, 95%

CAS: 26177-86-6 | C6H13Na2O9P | 306.11 g/mol

Supplier:  Thermo Scientific Chemicals J6631103

 View more versions of this product

Catalog No. AAJ6631103


Only null left
Explore available promotions

Description

Description

D-Fructose-6-phosphate disodium salt is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amidotransferase(s) and glucosamine-6P synthase(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-6-phosphate disodium salt is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amidotransferase(s) and glucosamine-6P synthase(s).

Solubility
Soluble in water (100 mg/ml).

Notes
Store at -20°C. Incompatible with heat and oxidizing agents.
Specifications

Specifications

D-Fructose-6-phosphate disodium salt
White
MFCD16661244
Soluble in water (100mg/ml).
[Na].[Na].OCC(=O)C(O)C(O)C(O)COP(O)(O)=O
306.11
304.1
Powder
26177-86-6
C6H13Na2O9P
1 g
VAPLWAFNCWDFQF-UHFFFAOYNA-N
sodium;[(2R,3R,4S)-2,3,4,6-tetrahydroxy-5-oxohexyl] dihydrogen phosphate
129893509
95%

Chemical Identifiers

26177-86-6
306.11
VAPLWAFNCWDFQF-UHFFFAOYNA-N
sodium;[(2R,3R,4S)-2,3,4,6-tetrahydroxy-5-oxohexyl] dihydrogen phosphate
C6H13Na2O9P
MFCD16661244
129893509
[Na].[Na].OCC(=O)C(O)C(O)C(O)COP(O)(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 247-505-4

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only