missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Fenchone, 97%

CAS: 4695-62-9 | C10H16O | 152.24 g/mol

$102.52 - $320.82

Chemical Identifiers

CAS 4695-62-9
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00070689
InChI Key LHXDLQBQYFFVNW-XCBNKYQSSA-N
Synonym +-fenchone, 1s-1,3,3-trimethylbicyclo 2.2.1 heptan-2-one, 1s-1,3,3-trimethyl-norbornan-2-one
PubChem CID 1201521
ChEBI CHEBI:165
IUPAC Name (1R,4S)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC(C2)(C1=O)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC366290250
View Documents Promotion Details
N/A 25 g Glass bottle
Each for $102.52
Only null left
 
AC366291000
View Documents Promotion Details
N/A 100 g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4695-62-9
152.24
LHXDLQBQYFFVNW-XCBNKYQSSA-N
1201521
(1R,4S)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
C10H16O
MFCD00070689
+-fenchone, 1s-1,3,3-trimethylbicyclo 2.2.1 heptan-2-one, 1s-1,3,3-trimethyl-norbornan-2-one
CHEBI:165
CC1(C2CCC(C2)(C1=O)C)C

Specifications

4695-62-9
Colorless to Yellow
63°C to 65°C (13.0 mmHg)
Authentic
C10H16O
MFCD00070689
0.95
+59.00 (20.00°C neat)
+-fenchone, 1s-1,3,3-trimethylbicyclo 2.2.1 heptan-2-one, 1s-1,3,3-trimethyl-norbornan-2-one
LHXDLQBQYFFVNW-XCBNKYQSSA-N
(1R,4S)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
1201521
152.24
Liquid
5°C to 7°C
0.9500g/mL
62°C
96% min. (GC)
1.4610 to 1.464
07, IV, 212
15, 4000
+ 59
Solubility in water: practically insoluble in water. Other solubilities: soluble in ethanol,ether,common organic solvents
CC1(C2CCC(C2)(C1=O)C)C
152.24
CHEBI:165
97%
D(+)-Fenchone, 97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 225-160-

SDS
Documents

Documents

RUO – Research Use Only