missing translation for 'onlineSavingsMsg'
Learn More

D-erythro-Sphingosine hydrochloride, 97%

Catalog No. AAH52427MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52427MC 100 mg
AAH52427ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52427MC Supplier Thermo Scientific Chemicals Supplier No. H52427MC

CAS: 2673-72-5 | C18H38ClNO2 | 335.96 g/mol

Inhibitor of protein kinase C and calmodulin-dependent enzymes, but may stimulate mast cells by activation of protein kinase C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2673-72-5
Molecular Formula C18H38ClNO2
Molecular Weight (g/mol) 335.96
MDL Number MFCD08436977
InChI Key YDIHJJLAPMAISR-UHFFFAOYNA-N
Synonym sphingosine hydrochloride, d-erythro-sphingosine hydrochloride, d-sphingosine hcl, trans-d-erythro-2-amino-4-octadecene-1,3-diol hydrochloride, 4-octadecene-1,3-diol,2-amino-,hydrochloride 1:1 , 2s,3r,4e, 4-octadecene-1,3-diol,2-amino-, hydrochloride 1:1 , 2s,3r,4e
PubChem CID 17998971
IUPAC Name (E,2S,3R)-2-aminooctadec-4-ene-1,3-diol;hydrochloride
SMILES Cl.CCCCCCCCCCCCCC=CC(O)C(N)CO

Specifications

Quantity 100 mg
Formula Weight 335.96
Percent Purity 97%
Chemical Name or Material D-erythro-Sphingosine hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only