accessibility menu, dialog, popup

UserName

CAS RN 2673-72-5

IUPAC Name: 2-aminooctadec-4-ene-1,3-diol hydrochloride
Synonyms: 2-aminooctadec-4-ene-1,3-diol hydrochloride
Molecular Weight (g/mol): 335.96
Molecular Formula: C18H38ClNO2
InChi Key: YDIHJJLAPMAISR-UHFFFAOYNA-N
SMILES: Cl.CCCCCCCCCCCCCC=CC(O)C(N)CO
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results