missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-erythro-Sphingosine 1-phosphate, 96%

Catalog No. AC328520010
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328520010 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC328520010 Supplier Thermo Scientific Chemicals Supplier No. 328520010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26993-30-6 | C18H38NO5P | 379.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26993-30-6
Molecular Formula C18H38NO5P
Molecular Weight (g/mol) 379.5
MDL Number MFCD00274457
InChI Key DUYSYHSSBDVJSM-KRWOKUGFSA-N
Synonym sphingosine 1-phosphate, sphingosine-1-phosphate, c18-sphingosine 1-phosphate, d-erythro-sphingosine-1-phosphate, sphing-4-enine 1-phosphate, d-erythro-sphingosine 1-phosphate, sphingosine 1-phosphic acid, s1p, sphing-4-enine-1-phosphate, 2s,3r,4e-2-amino-3-hydroxyoctadec-4-en-1-yl dihydrogen phosphate
PubChem CID 5283560
ChEBI CHEBI:37550
IUPAC Name [(E,2S,3R)-2-amino-3-hydroxyoctadec-4-enyl] dihydrogen phosphate
SMILES CCCCCCCCCCCCCC=CC(C(COP(=O)(O)O)N)O

Specifications

Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 95% min
Quantity 1 mg
Formula Weight 379.5
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material D-erythro-Sphingosine 1-phosphate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only