missing translation for 'onlineSavingsMsg'
Learn More

D-Epoxone, 98%

Catalog No. AAL1898906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1898906 5 g
AAL1898903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1898906 Supplier Thermo Scientific Chemicals Supplier No. L1898906
Only null left

CAS: 18422-53-2 | C12H18O6 | 258.27 g/mol

D-Epoxone is a chemical reagent used for enantioselective epoxidation of alkenes. It is involved in the synthesis of Irciniastatin (A and B), which are used as antitumor compounds. It is used as cytotoxic secondary metabolite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Epoxone is a chemical reagent used for enantioselective epoxidation of alkenes. It is involved in the synthesis of Irciniastatin (A and B), which are used as antitumor compounds. It is used as cytotoxic secondary metabolite.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 18422-53-2
Molecular Formula C12H18O6
Molecular Weight (g/mol) 258.27
MDL Number MFCD00063383
InChI Key IVWWFWFVSWOTLP-RDNCKJRISA-N
Synonym 3ar,4'r,7ar-2,2,2',2'-tetramethyldihydrospiro 1,3 dioxolo 4,5-c pyran-6,4'-1,3 dioxolan-7 7ah-one
PubChem CID 118990039
IUPAC Name (4S,7'aR)-2,2,2',2'-tetramethylspiro[1,3-dioxolane-4,6'-4,7a-dihydro-3aH-[1,3]dioxolo[4,5-c]pyran]-7'-one
SMILES CC1(OCC2(O1)C(=O)C3C(CO2)OC(O3)(C)C)C

Specifications

Melting Point 101°C to 103°C
Quantity 5 g
Beilstein 1319073
Solubility Information Soluble in chloroform and methanol.
Formula Weight 258.27
Percent Purity 98%
Chemical Name or Material D-Epoxone
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only