CAS RN 18422-53-2
IUPAC Name:
2,2,5',5'-tetramethyl-tetrahydro-2H-spiro[[1,3]dioxolo[4,5-c]pyran-6,2'-[1,4]dioxolan]-7-one
Synonyms:
2,2,5',5'-tetramethyl-dihydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-6,2'-[1,4]dioxolan]-7-one
Molecular Weight (g/mol):
258.27
Molecular Formula:
C12H18O6
InChi Key:
IVWWFWFVSWOTLP-UHFFFAOYNA-N
SMILES:
CC1(C)OC2COC3(COC(C)(C)O3)C(=O)C2O1
1
–
1
of
1
results
1
D-Epoxone, 98%
CAS: 18422-53-2 Molecular Formula: C12H18O6 Molecular Weight (g/mol): 258.27 MDL Number: MFCD00063383 InChI Key: IVWWFWFVSWOTLP-RDNCKJRISA-N Synonym: 3ar,4'r,7ar-2,2,2',2'-tetramethyldihydrospiro 1,3 dioxolo 4,5-c pyran-6,4'-1,3 dioxolan-7 7ah-one PubChem CID: 118990039 IUPAC Name: (4S,7'aR)-2,2,2',2'-tetramethylspiro[1,3-dioxolane-4,6'-4,7a-dihydro-3aH-[1,3]dioxolo[4,5-c]pyran]-7'-one SMILES: CC1(OCC2(O1)C(=O)C3C(CO2)OC(O3)(C)C)C