missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Cycloserine 98.0+%, TCI America™

Catalog No. C11891G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11891G 1 g
C11895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11891G Supplier TCI America Supplier No. C11891G
Only null left

Chemical Identifiers

CAS 68-41-7
Molecular Formula C3H6N2O2
Molecular Weight (g/mol) 102.093
MDL Number MFCD00005353
InChI Key DYDCUQKUCUHJBH-UWTATZPHSA-N
Synonym d-cycloserine, cycloserine, seromycin, orientomycin, oxamycin, cyclorin, cyclo-d-serine, cicloserina, farmiserina, miroseryn
PubChem CID 6234
ChEBI CHEBI:40009
IUPAC Name (4R)-4-amino-1,2-oxazolidin-3-one
SMILES C1C(C(=O)NO1)N

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 1 g
Formula Weight 102.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Cycloserine
RTECSNumber NY2975000
TSCA No
Recommended Storage Refrigerator