missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Cellopentaose, 95%, Thermo Scientific Chemicals

$217.14 - $439.56

Chemical Identifiers

CAS 2240-27-9
MDL Number MFCD09750881
Synonym Cellopentaose
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67561MCR
View Documents
Thermo Scientific Chemicals
J67561MCR
1 mg
Each for $217.14
Only null left
 
AAJ67561LB0
View Documents
Thermo Scientific Chemicals
J67561LB0
5 mg
Each for $439.56
Only null left
 
Description

Description

Used by cellulosic bacteria as an energy source. Used to identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and --glucosidase(s). Can also be used in saccharification and ethanol fermentation research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used by cellulosic bacteria as an energy source
  • Used to identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and _x001A_-glucosidase(s)
  • Can also be used in saccharification and ethano
Specifications

Chemical Identifiers

2240-27-9
MFCD09750881
Cellopentaose

Specifications

2240-27-9
MFCD09750881
79497
Cellopentaose
95%
D-(+)-Cellopentaose
White
1 mg
Hygroscopic
828.72
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only