Learn More
D-(+)-Cellopentaose, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J67561MCR
Description
Used by cellulosic bacteria as an energy source. Used to identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and --glucosidase(s). Can also be used in saccharification and ethanol fermentation research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Used by cellulosic bacteria as an energy source
- Used to identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and _x001A_-glucosidase(s)
- Can also be used in saccharification and ethano
Specifications
| D-(+)-Cellopentaose | |
| White | |
| 1 mg | |
| Hygroscopic | |
| 828.72 | |
| Powder |
| 2240-27-9 | |
| MFCD09750881 | |
| 79497 | |
| Cellopentaose | |
| 95% |
Chemical Identifiers
| 2240-27-9 | |
| MFCD09750881 | |
| Cellopentaose |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only