missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Cellohexaose, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J676108PL

 View more versions of this product

Catalog No. AAJ676108PL


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D-(+)-Cellohexaose
White
2 mg
Cellohexaose
95%
2478-35-5
MFCD00466913
Hygroscopic
990.86
Powder

Chemical Identifiers

2478-35-5
MFCD00466913
Cellohexaose
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only