CAS RN 2478-35-5
CAS RN 2478-35-5
IUPAC Name:
(2R,3R,4R,5R)-4-{[(2S,3R,4R,5S,6R)-5-{[(2S,3R,4R,5S,6R)-5-{[(2S,3R,4R,5S,6R)-5-{[(2S,3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]oxy}-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2,3,5,6-tetrahydroxyhexanal
Synonyms:
(2R,3R,4R,5R)-4-{[(2S,3R,4R,5S,6R)-5-{[(2S,3R,4R,5S,6R)-5-{[(2S,3R,4R,5S,6R)-5-{[(2S,3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]oxy}-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2,3,5,6-tetrahydroxyhexanal
Molecular Weight (g/mol):
990.86
Molecular Formula:
C36H62O31
InChi Key:
DJMVHSOAUQHPSN-VXDFXQCISA-N
SMILES:
OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O[C@@H]4O[C@H](CO)[C@@H](O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)[C@H](O)[C@H]4O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O
1
–
1
of
1
results
1
D-(+)-Cellohexaose, 95%, Thermo Scientific Chemicals
CAS: 2478-35-5 MDL Number: MFCD00466913 Synonym: Cellohexaose