missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Cellobiose octaacetate

Catalog No. p-7044933
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0878006 5 g
AAL0878014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0878006 Supplier Thermo Scientific Chemicals Supplier No. L0878006

CAS: 5346-90-7 | C28H38O19 | 678.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5346-90-7
Molecular Formula C28H38O19
Molecular Weight (g/mol) 678.59
MDL Number MFCD00009600,MFCD00079432,MFCD00009600,MFCD00078092,MFCD00009600,MFCD00009600
InChI Key WOTQVEKSRLZRSX-UHFFFAOYNA-N
Synonym d-cellobiose octaacetate
PubChem CID 126963588
IUPAC Name [(2R,3R,4R,5R,6S)-4,5,6-triacetyloxy-3-[(2S,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC1OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 224°C to 231°C
Quantity 5 g
Beilstein 79280
Merck Index 14,1961
Optical Rotation +40° (c=1 in Chloroform)
Formula Weight 678.59
Chemical Name or Material D-Cellobiose octaacetate
EINECSNumber 226-304-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only