accessibility menu, dialog, popup

UserName

H3COOOOCH3OOCH3OOCH3OOOOCH3OOCH3OOCH3OOCH3O

CAS RN 5346-90-7

IUPAC Name: [4,5,6-tris(acetyloxy)-3-{[3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxan-2-yl]methyl acetate
Synonyms: [4,5,6-tris(acetyloxy)-3-{[3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 678.59
Molecular Formula: C28H38O19
InChi Key: WOTQVEKSRLZRSX-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC1OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)