missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D(+)-Cellobiose, 98%
CAS: 528-50-7 | C12H22O11 | 342.30 g/mol
Supplier: Thermo Scientific Chemicals 108460250
| Quantity | 25 g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C12H22O11 | |
| MFCD00136034 | |
| d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose | |
| (3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
Specifications
| D(+)-Cellobiose | |
| 239°C | |
| Authentic | |
| Glass bottle | |
| 25 g | |
| 31, 380 | |
| + 34 | |
| (5% in water) Clear colorless | |
| OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O | |
| 342.30 | |
| 342.29 | |
| Fine Crystalline Powder |
| 528-50-7 | |
| White | |
| 97.5% min. (ELSD) (HPLC) | |
| C12H22O11 | |
| MFCD00136034 | |
| 15, 1963 | |
| d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose | |
| GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
| (3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol | |
| 57370084 | |
| 98% |
Safety and Handling
EINECSNumber : 208-436-5