missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Cellobiose, 98%

Catalog No. p-3735816
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108460050 5 g Glass bottle
AC108460250 25 g Glass bottle
AC108461000 100 g Glass bottle
AC108465000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC108460050 Supplier Thermo Scientific Chemicals Supplier No. 108460050

CAS: 528-50-7 | C12H22O11 | 342.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 528-50-7
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00136034
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
PubChem CID 57370084
IUPAC Name (3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Melting Point 239°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 31, 380
Merck Index 15, 1963
Specific Rotation + 34
Solubility Information (5% in water) Clear colorless
Formula Weight 342.29
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material D(+)-Cellobiose
Show More Show Less
EINECSNumber 208-436-5

RUO – Research Use Only