Learn More
Thermo Scientific Chemicals D-(+)-Cellobiose, 98+%
CAS: 528-50-7 | C12H22O11 | 342.30 g/mol
Supplier: Thermo Scientific Chemicals A1455306
Description
D-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.
Solubility
Soluble in water (1 g/8ml). Insoluble in ether and ethanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| D-(+)-Cellobiose | |
| 234°C (decomposition) | |
| C12H22O11 | |
| 5 g | |
| 14,1961 | |
| Soluble in water (1g/8ml). Insoluble in ether and ethanol. | |
| +34° (c=10 in Water, 15h) | |
| (3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol | |
| 57370084 | |
| ≥98% |
| 528-50-7 | |
| ≥98% | |
| MFCD00136034 | |
| 93795 | |
| d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose | |
| GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
| OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O | |
| 342.30 | |
| 342.3 |
Chemical Identifiers
| 528-50-7 | |
| 342.30 | |
| GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
| 57370084 |
| C12H22O11 | |
| MFCD00136034 | |
| d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose | |
| OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O |
Safety and Handling
EINECSNumber : 208-436-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only