missing translation for 'onlineSavingsMsg'
Learn More

D and C Yellow No. 10, Spectrum™ Chemical

DC200, 8004-92-0, C18H9NO8S2Na

Supplier:  Spectrum Chemical Mfg Cor DC200500GM

 View more versions of this product

Catalog No. 18601991


Only null left

Description

Description

Spectrum™ Chemical D and C Yellow No. 10
Specifications

Specifications

D and C Yellow No. 10
100%
MFCD00080727
[Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2N=C(C=CC2=C1)C1C(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O
477.37
Reagent
Yellow
8004-92-0
C18H9NNa2O8S2
FZUOVNMHEAPVBW-UHFFFAOYSA-L
disodium 2-(1,3-dioxo-2,3-dihydro-1H-inden-2-yl)quinoline-6,8-disulfonate
85%
Amber Glass Bottle
500 g

Chemical Identifiers

8004-92-0
477.37
FZUOVNMHEAPVBW-UHFFFAOYSA-L
[Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2N=C(C=CC2=C1)C1C(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O
C18H9NNa2O8S2
MFCD00080727
disodium 2-(1,3-dioxo-2,3-dihydro-1H-inden-2-yl)quinoline-6,8-disulfonate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions