missing translation for 'onlineSavingsMsg'
Learn More

D and C Yellow No. 10, Spectrum™ Chemical

Catalog No. p-7157337
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601989 25 g Amber Glass Bottle
18601990 100 g Amber Glass Bottle
18601991 500 g Amber Glass Bottle
18601992 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601989 Supplier Spectrum Chemical Mfg Cor Supplier No. DC20025GM
Only null left

DC200, 8004-92-0, C18H9NO8S2Na

Spectrum™ Chemical D and C Yellow No. 10

Chemical Identifiers

CAS 8004-92-0
Molecular Formula C18H9NNa2O8S2
Molecular Weight (g/mol) 477.37
MDL Number MFCD00080727
InChI Key FZUOVNMHEAPVBW-UHFFFAOYSA-L
IUPAC Name disodium 2-(1,3-dioxo-2,3-dihydro-1H-inden-2-yl)quinoline-6,8-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2N=C(C=CC2=C1)C1C(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O

Specifications

CAS Max % 100%
Percent Purity 85%
Grade Reagent
Packaging Amber Glass Bottle
Color Yellow
Quantity 25 g
Chemical Name or Material D and C Yellow No. 10