missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D(+)-Biotin, 98%
CAS: 58-85-5 | C10H16N2O3S | 244.31 g/mol
Supplier: Thermo Scientific Chemicals 230090050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| D(+)-Biotin | |
| 231.0°C to 233.0°C | |
| Glass bottle | |
| MFCD00005541 | |
| 15,1236 | |
| + 89.00 | |
| Solubility in water: 0.22g/L (25°C). Other solubilities: more soluble in hot water and dil. alkali,0.8g/L alcohol,insoluble in most other common organic solvents | |
| OC(=O)CCCCC1SCC2NC(=O)NC12 | |
| 244.31 | |
| CHEBI:15956 | |
| 98% |
| 58-85-5 | |
| 98% | |
| C10H16N2O3S | |
| 5 g | |
| + 89.00 (20.00°C c=1,0.1N NaOH) | |
| biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin | |
| YBJHBAHKTGYVGT-UHFFFAOYNA-N | |
| 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid | |
| 171548 | |
| 244.31 |
Identifiants chimiques
| 58-85-5 | |
| 244.31 | |
| YBJHBAHKTGYVGT-UHFFFAOYNA-N | |
| 171548 | |
| 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid |
| C10H16N2O3S | |
| MFCD00005541 | |
| biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin | |
| CHEBI:15956 | |
| OC(=O)CCCCC1SCC2NC(=O)NC12 |
Sécurité et manipulation
missing translation for 'einecsNumber' : 200-399-3
RUO – Research Use Only