missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Biotin, 98%

Catalog No. p-3735618
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC230095000 500 mg Glass bottle
AC230090010 1 g Glass bottle
AC230090050 5 g Glass bottle
AC230090250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC230095000 Supplier Thermo Scientific Chemicals Supplier No. 230095000

CAS: 58-85-5 | C10H16N2O3S | 244.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
Synonym biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
PubChem CID 171548
ChEBI CHEBI:15956
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

Melting Point 231.0°C to 233.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 500 mg
Merck Index 15,1236
Specific Rotation + 89.00
Solubility Information Solubility in water: 0.22g/L (25°C). Other solubilities: more soluble in hot water and dil. alkali,0.8g/L alcohol,insoluble in most other common organic solvents
Formula Weight 244.31
Percent Purity 98%
Chemical Name or Material D(+)-Biotin
EINECSNumber 200-399-3

RUO – Research Use Only