missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D(+)-Biotin, 98%
CAS: 58-85-5 | C10H16N2O3S | 244.31 g/mol
Supplier: Thermo Scientific Chemicals 230090010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| D(+)-Biotin | |
| 231.0°C to 233.0°C | |
| Glass Bottle | |
| MFCD00005541 | |
| 15,1236 | |
| biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin | |
| YBJHBAHKTGYVGT-UHFFFAOYNA-N | |
| 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid | |
| 171548 | |
| 244.31 |
| 58-85-5 | |
| 98% | |
| C10H16N2O3S | |
| 1 g | |
| + 89.00 | |
| Solubility in water: 0.22g/L (25°C). Other solubilities: more soluble in hot water and dil. alkali,0.8g/L alcohol,insoluble in most other common organic solvents | |
| OC(=O)CCCCC1SCC2NC(=O)NC12 | |
| 244.31 | |
| CHEBI:15956 | |
| 98% |
Chemical Identifiers
| 58-85-5 | |
| 244.31 | |
| YBJHBAHKTGYVGT-UHFFFAOYNA-N | |
| 171548 | |
| 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid |
| C10H16N2O3S | |
| MFCD00005541 | |
| biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin | |
| CHEBI:15956 | |
| OC(=O)CCCCC1SCC2NC(=O)NC12 |
Safety and Handling
EINECSNumber : 200-399-3
RUO – Research Use Only